missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-7-methylthieno[3,2-d]pyrimidine, 97%, Thermo Scientific™

Product Code. 10543032
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-21-0
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.64
MDL Number MFCD00205202
InChI Key IYJDOVYAFDVIDB-UHFFFAOYSA-N
PubChem CID 2777586
IUPAC Name 4-chloro-7-methylthieno[3,2-d]pyrimidine
SMILES CC1=CSC2=C(Cl)N=CN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4-chloro-7-methylthieno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 4-chloro-7-methyl, 4-chloro-7-methylthiopheno 3,2-d pyrimidine, thieno 3,2-d pyrimidine,4-chloro-7-methyl, pubchem23125, 4-chloro-7-methyl thieno 3,2-d pyrimidine, 4-chloro-7-methyl-thieno 3,2-d pyrimidine, 4-chloranyl-7-methyl-thieno 3,2-d pyrimidine, 4-chloro-7-methylthieno 2,3-d pyrimidine
Formula Weight 184.65
Percent Purity 97%
Chemical Name or Material 4-Chloro-7-methylthieno[3, 2-d]pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.