missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-N-(2-fluorobenzyl)aniline, 97%, Thermo Scientific™ Product Code.: 15482728

4-Chloro-N-(2-fluorobenzyl)aniline, 97%, Thermo Scientific™

Product Code. 15482728
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15482728

Brand: Thermo Scientific Alfa Aesar H56275.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723753-84-2
Molecular Formula C13H11ClFN
Molecular Weight (g/mol) 235.69
MDL Number MFCD05740323
InChI Key QCLJXZCWBRKJHD-UHFFFAOYSA-N
Synonym 4-chloro-n-2-fluorophenyl methyl aniline, n-4-chlorophenyl-n-2-fluorobenzyl amine, 4-chloro-n-2-fluorobenzyl aniline
PubChem CID 834588
IUPAC Name 4-chloro-N-[(2-fluorophenyl)methyl]aniline
SMILES FC1=CC=CC=C1CNC1=CC=C(Cl)C=C1

Specifications

Quantity 250 mg
Formula Weight 235.69
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(2-fluorobenzyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-N-(2-fluorobenzyl)aniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.