missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-cyclopentyl-3-nitrobenzamide, 97%, Thermo Scientific™

£102.00 - £325.00

Chemical Identifiers

CAS 339059-83-5
Molecular Formula C12H13ClN2O3
Molecular Weight (g/mol) 268.697
MDL Number MFCD00738373
InChI Key VZZDMSAFALTDME-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl-n-cyclopentylcarboxamide
PubChem CID 767775
IUPAC Name 4-chloro-N-cyclopentyl-3-nitrobenzamide
SMILES C1CCC(C1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485968
View Documents
Thermo Scientific Alfa Aesar
H59595.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15475968
View Documents
Thermo Scientific Alfa Aesar
H59595.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

339059-83-5
268.697
VZZDMSAFALTDME-UHFFFAOYSA-N
767775
C1CCC(C1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
C12H13ClN2O3
MFCD00738373
4-chloro-3-nitrophenyl-n-cyclopentylcarboxamide
4-chloro-N-cyclopentyl-3-nitrobenzamide

Specifications

339059-83-5
MFCD00738373
4-chloro-3-nitrophenyl-n-cyclopentylcarboxamide
C1CCC(C1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
268.697
268.7
4-Chloro-N-cyclopentyl-3-nitrobenzamide
C12H13ClN2O3
20162022
VZZDMSAFALTDME-UHFFFAOYSA-N
4-chloro-N-cyclopentyl-3-nitrobenzamide
767775
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.