missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-N'-hydroxybenzenecarboximidamide, 95%, Thermo Scientific™ Product Code.: 10462474

4-Chloro-N'-hydroxybenzenecarboximidamide, 95%, Thermo Scientific™

Product Code. 10462474
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10462474

Brand: Thermo Scientific Maybridge SPB06960CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5033-28-3
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00029674
InChI Key QBGONPQFBDUVPG-UHFFFAOYSA-N
Synonym z-4-chloro-n'-hydroxybenzamidine, 4-chloro-n-hydroxy-benzamidine, 4-chloro-n-hydroxybenzenecarboximidamide, 4-chlorobenzamide oxime, z-4-chloro-n'-hydroxybenzimidamide, 4-chloro-n'-hydroxybenzene-1-carboximidamide, z-4-chloro-n'-hydroxybenzene-1-carboximidamide, 4-chloro-n'-hydroxybenzimidamide, 4-chloro-benzamidoxime, 4-chlorophenyl hydroxyimino methylamine
PubChem CID 9561067
IUPAC Name (Z)-4-chloro-N'-hydroxybenzene-1-carboximidamide
SMILES N\C(=N/O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 95.0
Quantity 250 mg
Assay Percent Range 95%
Linear Formula ClC6H4C(=NOH)NH2
Formula Weight 170.6
Percent Purity 95%
Chemical Name or Material 4-Chloro-N'-hydroxybenzenecarboximidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-N'-hydroxybenzenecarboximidamide, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.