missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Chloro-N-methylbenzylamine, 95%
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 77°C to 78°C (1 mmHg) |
Quantity | 5 g |
UN Number | UN2735 |
Formula Weight | 155.62 |
Percent Purity | 95% |
Chemical Name or Material | 4-Chloro-N-methylbenzylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'