Learn More
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
£25.30 - £203.00
Chemical Identifiers
CAS | 99-91-2 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.59 |
MDL Number | MFCD00000624 |
InChI Key | BUZYGTVTZYSBCU-UHFFFAOYSA-N |
Synonym | 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene |
PubChem CID | 7467 |
ChEBI | CHEBI:27538 |
IUPAC Name | 1-(4-chlorophenyl)ethanone |
SMILES | CC(=O)C1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10284370
|
Thermo Scientific Acros
108540050 |
5 g | Glass bottle |
£25.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10375880
|
Thermo Scientific
108542500 |
250 g | Glass bottle |
£74.30
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10257263
|
Thermo Scientific Acros
108540010 |
1 kg | Glass bottle |
£203.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-91-2 | |
154.59 | |
BUZYGTVTZYSBCU-UHFFFAOYSA-N | |
7467 | |
1-(4-chlorophenyl)ethanone |
C8H7ClO | |
MFCD00000624 | |
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene | |
CHEBI:27538 | |
CC(=O)C1=CC=C(Cl)C=C1 |
Specifications
99-91-2 | |
100.0 | |
Colorless to Yellow | |
232.0°C | |
Authentic | |
C8H7ClO | |
1.5539 to 1.5559 | |
07,281 | |
1.19 | |
Solubility in water: 111 mg/l (25°C). | |
CC(=O)C1=CC=C(Cl)C=C1 | |
154.59 | |
CHEBI:27538 | |
98+% | |
4'-Chloroacetophenone, 97% |
96.0 | |
18.0°C | |
1.1900g/mL | |
126°C | |
96% min. (GC) | |
ClC6H4COCH3 | |
MFCD00000624 | |
15,2116 | |
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene | |
BUZYGTVTZYSBCU-UHFFFAOYSA-N | |
1-(4-chlorophenyl)ethanone | |
7467 | |
154.6 | |
Liquid After Melting |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-800-7
missing translation for 'rtecsNumber' : KM5600000
missing translation for 'tsca' : TSCA
- 4'-Chloroacetophenone
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only