missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
Solubility
Soluble in water (2.6 g/Liter at 20°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.
Solubility
Soluble in water (2.6 g/Liter at 20°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
Specifications
Melting Point | 67°C to 72°C |
Density | 1.43 |
Boiling Point | 232°C |
Quantity | 250 g |
UN Number | UN2018 |
Beilstein | 471359 |
Merck Index | 14,2118 |
Solubility Information | Soluble in water (2.6g/Liter at 20°C ). |
Formula Weight | 127.57 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'