missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzene-1-carboximidamide hydrochloride, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Linear Formula | ClC6H4C(=NH)NH2.HCl |
Formula Weight | 191.06 |
Percent Purity | 95% |
Quantity | 1 g |
Packaging | Amber Glass Bottle |
Chemical Name or Material | 4-Chlorobenzene-1-carboximidamide hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction