missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Chlorobenzenesulfonamide, 98%
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Melting Point | 144°C to 148°C |
Quantity | 100 g |
Beilstein | 1308891 |
Formula Weight | 191.64 |
Percent Purity | 98% |
Chemical Name or Material | 4-Chlorobenzenesulfonamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'