missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chlorobenzenesulfonamide, 98%

Product Code. 11422748
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 98-64-6
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.63
MDL Number MFCD00007936
InChI Key HHHDJHHNEURCNV-UHFFFAOYSA-N
Synonym p-chlorobenzenesulfonamide, benzenesulfonamide, 4-chloro, p-chlorobenzenesulfamide, 4-chlorophenylsulfonamide, p-chlorophenylsulfonamide, 4-chlorobenzenesulphonamide, benzenesulfonamide, p-chloro, usaf ma-3, 4-chlorobenzene-1-sulfonamide, 4-chloro-benzenesulfonamide
PubChem CID 66824
IUPAC Name 4-chlorobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 144°C to 148°C
Quantity 500 g
Beilstein 1308891
Formula Weight 191.64
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'