missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydryl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11449817
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-83-8
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000856
InChI Key ALKWTKGPKKAZMN-UHFFFAOYSA-N
Synonym 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene
PubChem CID 241584
IUPAC Name 1-chloro-4-[chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl

Specifications

Density 1.239
Boiling Point 159°C to 160°C (2 mmHg)
Flash Point 194°C (381°F)
Refractive Index 1.603
Quantity 25 g
UN Number UN3265
Beilstein 2050236
Sensitivity Moisture sensitive
Formula Weight 237.13
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzhydryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'