Learn More
4-Chlorobenzotrifluoride, 98%, Thermo Scientific Chemicals
£15.30 - £137.00
Chemical Identifiers
CAS | 98-56-6 |
---|---|
Molecular Formula | C7H4ClF3 |
Molecular Weight (g/mol) | 180.56 |
MDL Number | MFCD00000627 |
InChI Key | QULYNCCPRWKEMF-UHFFFAOYSA-N |
Synonym | 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene |
PubChem CID | 7394 |
IUPAC Name | 1-chloro-4-(trifluoromethyl)benzene |
SMILES | C1=CC(=CC=C1C(F)(F)F)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10224230
|
Thermo Scientific Acros
108730250 |
25 mL | Glass bottle |
£15.30
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091020
|
Thermo Scientific Acros
108732500 |
250 mL | Glass bottle |
£43.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10035913
|
Thermo Scientific Acros
108730010 |
1 L | Glass bottle |
£137.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-56-6 | |
180.56 | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
7394 | |
C1=CC(=CC=C1C(F)(F)F)Cl |
C7H4ClF3 | |
MFCD00000627 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
1-chloro-4-(trifluoromethyl)benzene |
Specifications
98-56-6 | |
100.0 | |
Colorless | |
136.0°C to 138.0°C | |
Authentic | |
C7H4ClF3 | |
1.4453 to 1.4473 | |
15, 2127 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
C1=CC(=CC=C1C(F)(F)F)Cl | |
180.56 | |
0.67 mPa.s (38°C) | |
98% | |
4-Chlorobenzotrifluoride, 98% |
97.5 | |
-36.0°C | |
1.3500g/mL | |
47°C | |
98% | |
ClC6H4CF3 | |
MFCD00000627 | |
1.35 | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
1-chloro-4-(trifluoromethyl)benzene | |
7394 | |
180.56 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-681-1
missing translation for 'rtecsNumber' : XS9145000
missing translation for 'tsca' : TSCA
- 4-Chlorobenzotrifluoride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only