Learn More
4-Chlorobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Description
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
Specifications
Specifications
Melting Point | -36°C |
Density | 1.346 |
Boiling Point | 136°C to 138°C |
Flash Point | 47°C (116°F) |
Refractive Index | 1.446 |
Quantity | 500 g |
UN Number | UN2234 |
Beilstein | 510203 |
Merck Index | 14,2126 |
Solubility Information | Miscible with chloroform. Slightly miscible with water. |
Show More |
RUO – Research Use Only