Learn More
4-Chlorobenzotrifluoride, 98+%, Thermo Scientific Chemicals
£15.30 - £205.00
Chemical Identifiers
CAS | 98-56-6 |
---|---|
Molecular Formula | C7H4ClF3 |
Molecular Weight (g/mol) | 180.554 |
MDL Number | MFCD00000627 |
InChI Key | QULYNCCPRWKEMF-UHFFFAOYSA-N |
Synonym | 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene |
PubChem CID | 7394 |
IUPAC Name | 1-chloro-4-(trifluoromethyl)benzene |
SMILES | C1=CC(=CC=C1C(F)(F)F)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11442138
|
Thermo Scientific Alfa Aesar
A15154.22 |
100 g |
£15.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452138
|
Thermo Scientific Alfa Aesar
A15154.36 |
500 g |
£58.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462138
|
Thermo Scientific Alfa Aesar
A15154.0E |
2500 g |
£205.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
Chemical Identifiers
98-56-6 | |
180.554 | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
7394 | |
C1=CC(=CC=C1C(F)(F)F)Cl |
C7H4ClF3 | |
MFCD00000627 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
1-chloro-4-(trifluoromethyl)benzene |
Specifications
98-56-6 | |
1.346 | |
47°C (116°F) | |
1.446 | |
UN2234 | |
14,2126 | |
Miscible with chloroform. Slightly miscible with water. | |
C1=CC(=CC=C1C(F)(F)F)Cl | |
180.554 | |
180.56 | |
4-Chlorobenzotrifluoride |
-36°C | |
136°C to 138°C | |
C7H4ClF3 | |
MFCD00000627 | |
510203 | |
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
QULYNCCPRWKEMF-UHFFFAOYSA-N | |
1-chloro-4-(trifluoromethyl)benzene | |
7394 | |
≥98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES
missing translation for 'einecsNumber' : 202-681-1
missing translation for 'rtecsNumber' : XS9145000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only