missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoylacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11469503
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4640-66-8
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051625
InChI Key JYOUFPNYTOFCSJ-UHFFFAOYSA-N
Synonym 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
PubChem CID 138308
IUPAC Name 3-(4-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC(=CC=C1C(=O)CC#N)Cl

Specifications

Melting Point 128°C to 132°C
Quantity 25 g
Beilstein 743368
Solubility Information Soluble in Dichloromethane,Ether,Ethyl Acetate and Methanol
Formula Weight 179.61
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzoylacetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'