missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl mercaptan, 98%, Thermo Scientific™

Product Code. 11488707
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6258-66-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00004870
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
PubChem CID 80409
IUPAC Name (4-chlorophenyl)methanethiol
SMILES SCC1=CC=C(Cl)C=C1

Specifications

Melting Point 19°C to 20°C
Density 1.202
Boiling Point 234°C
Quantity 100 g
Odor Stench
Refractive Index 1.5895
UN Number UN2810
Beilstein 606496
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzyl mercaptan
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'