missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15415375
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzylzinc chloride used as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a
TRUSTED_SUSTAINABILITY

Specifications

CAS 149923-10-4
Density 0.96
Quantity 50 mL
Molecular Formula C7H6Cl2Zn
MDL Number MFCD01319882
UN Number UN3399
Synonym 4-chlorobenzylzinc chloride solution, 4-chlorobenzylzinc chloride, chloro 4-chlorophenyl methyl zinc, 4-chlorobenzylzinc chloride solution, 0.5 m in thf, 4-chlorobenzylzinc chloride 0.5 m in tetrahydrofuran, 4-chlorobenzylzinc chloride, 0.5m in thf, packaged under arg, 4-chlorobenzylzinc chloride,0.5m soluti onin tetrahydrofuran, 4-chlorobenzylzinc chloride, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key GIUOJMHOGPURPS-UHFFFAOYSA-M
SMILES Cl[Zn+].[CH2-]C1=CC=C(Cl)C=C1
IUPAC Name 1-chloro-4-methanidylbenzene;chlorozinc(1+)
Molecular Weight (g/mol) 226.41
PubChem CID 3451123
Formula Weight 226.41
Chemical Name or Material 4-Chlorobenzylzinc chloride, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'