missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobutyronitrile, 97%, Thermo Scientific Chemicals

£17.50 - £188.94

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10255671
View Documents
Thermo Scientific Acros
329940050
5 g Glass bottle
£17.50
5g
Please to purchase this item. Need a web account? Register with us today!
10402661
View Documents
Thermo Scientific Acros
329941000
100 g Glass bottle
£45.12
100g
Please to purchase this item. Need a web account? Register with us today!
10167763
View Documents
Thermo Scientific Acros
329945000
500 g Glass bottle
£188.94
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

628-20-6
103.55
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
12336
ClCCCC#N
C4H6ClN
MFCD00001972
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
4-chlorobutanenitrile

Specifications

628-20-6
100.0
1.0800g/mL
85°C
96% min. (GC)
1.4420 to 1.444
02, 278
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutanenitrile
12336
97%
4-chlorobutyronitrile
96.0
Colorless to Yellow
195°C to 197°C
Authentic
C4H6ClN
MFCD00001972
1.08
Solubility in water: immiscible
ClCCCC#N
103.55
103.55
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 211-031-6

Product Identifier
  • 4-Chlorobutyronitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only