Learn More
4-Chlorobutyryl chloride, 98%, Thermo Scientific Chemicals
£23.20 - £59.70
Chemical Identifiers
CAS | 4635-59-0 |
---|---|
Molecular Formula | C4H6Cl2O |
Molecular Weight (g/mol) | 141 |
MDL Number | MFCD00000754 |
InChI Key | CDIIZULDSLKBKV-UHFFFAOYSA-N |
Synonym | 4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro |
PubChem CID | 78370 |
IUPAC Name | 4-chlorobutanoyl chloride |
SMILES | C(CC(=O)Cl)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10244420
|
Thermo Scientific Acros
108902500 |
250 mL |
£23.20
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10269693
|
Thermo Scientific Acros
108900010 |
1 L |
£59.70
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4635-59-0 | |
141 | |
CDIIZULDSLKBKV-UHFFFAOYSA-N | |
78370 | |
C(CC(=O)Cl)CCl |
C4H6Cl2O | |
MFCD00000754 | |
4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro | |
4-chlorobutanoyl chloride |
Specifications
4635-59-0 | |
100.0 | |
Colorless to Yellow | |
173.0°C to 174.0°C | |
Authentic | |
Glass bottle | |
Cl(CH2)3COCl | |
MFCD00000754 | |
1.25 | |
Solubility in water: reacts. | |
C(CC(=O)Cl)CCl | |
141 | |
141 | |
Liquid |
97.5 | |
-47.0°C | |
1.2500g/mL | |
85°C | |
97.5% min. (GC) | |
C4H6Cl2O | |
1.4605 to 1.4625 | |
02, 278 | |
4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro | |
CDIIZULDSLKBKV-UHFFFAOYSA-N | |
4-chlorobutanoyl chloride | |
78370 | |
98% | |
4-Chlorobutyryl chloride, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear eye protection/face protecti
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 225-059-1
missing translation for 'rtecsNumber' : EM1406000
missing translation for 'tsca' : TSCA
- 4-Chlorobutyryl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only