missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocinnamic acid, 99%, predominantly trans

Product Code. 10617062 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10617062

Brand: Thermo Scientific Acros 108940250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1615-02-7 | C9H7ClO2 | 182.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1615-02-7
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.60
MDL Number MFCD00004396
InChI Key GXLIFJYFGMHYDY-ZZXKWVIFSA-N
Synonym 4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
PubChem CID 637797
ChEBI CHEBI:61116
IUPAC Name (E)-3-(4-chlorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Cl)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 246°C to 250°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClC6H4CH=CHCO2H
Quantity 25 g
Beilstein 09, 594
Formula Weight 182.61
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Chlorocinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorocinnamic acid, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.