missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)benzyl alcohol, 97%, Thermo Scientific Chemicals
£43.00 - £143.00
Chemical Identifiers
CAS | 16473-35-1 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.61 |
InChI Key | OGALXJIOJZXBBP-UHFFFAOYSA-N |
Synonym | 4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride |
PubChem CID | 5107660 |
IUPAC Name | [4-(chloromethyl)phenyl]methanol |
SMILES | C1=CC(=CC=C1CO)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11994651
|
Thermo Scientific Acros
438920010 |
1 g |
£43.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11904661
|
Thermo Scientific Acros
438920050 |
5 g |
£143.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16473-35-1 | |
156.61 | |
4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride | |
[4-(chloromethyl)phenyl]methanol |
C8H9ClO | |
OGALXJIOJZXBBP-UHFFFAOYSA-N | |
5107660 | |
C1=CC(=CC=C1CO)CCl |
Specifications
16473-35-1 | |
Authentic | |
Glass bottle | |
ClCH2C6H4CH2OH | |
OGALXJIOJZXBBP-UHFFFAOYSA-N | |
[4-(chloromethyl)phenyl]methanol | |
5107660 | |
97% |
58.0°C to 60.0°C | |
96% min. (GC) | |
C8H9ClO | |
4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride | |
C1=CC(=CC=C1CO)CCl | |
156.61 | |
156.61 | |
4-(Chloromethyl)benzyl alcohol |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-(Chloromethyl)benzyl alcohol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only