missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Chlorophenylethynyl)trimethylsilane, 97%, Thermo Scientific™
£22.80 - £260.00
Chemical Identifiers
CAS | 78704-49-1 |
---|---|
Molecular Formula | C11H13ClSi |
Molecular Weight (g/mol) | 208.76 |
MDL Number | MFCD03427267 |
InChI Key | HVIHXUCWKNRJTC-UHFFFAOYSA-N |
Synonym | 4-chlorophenylethynyl trimethylsilane, 4-chlorophenyl ethynyl trimethylsilane, 2-4-chlorophenyl ethynyl trimethylsilane, 2 4-chlorophenyl-1-trimethylsilylethyne, 2-4-chlorophenyl ethynyl-trimethylsilane, acmc-1bg9d, chloro-4-trimethylsilylethynylbenzene, trimethyl 4-chlorophenylethynyl silane, 2 4'-chlorophenyl-1-trimethylsilylethyne |
PubChem CID | 3337298 |
IUPAC Name | 2-(4-chlorophenyl)ethynyl-trimethylsilane |
SMILES | C[Si](C)(C)C#CC1=CC=C(C=C1)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461278
|
Thermo Scientific Alfa Aesar
H55129.03 |
1 g |
£22.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15471278
|
Thermo Scientific Alfa Aesar
H55129.06 |
5 g | |||||||
15481278
|
Thermo Scientific Alfa Aesar
H55129.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
78704-49-1 | |
208.76 | |
HVIHXUCWKNRJTC-UHFFFAOYSA-N | |
3337298 | |
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl |
C11H13ClSi | |
MFCD03427267 | |
4-chlorophenylethynyl trimethylsilane, 4-chlorophenyl ethynyl trimethylsilane, 2-4-chlorophenyl ethynyl trimethylsilane, 2 4-chlorophenyl-1-trimethylsilylethyne, 2-4-chlorophenyl ethynyl-trimethylsilane, acmc-1bg9d, chloro-4-trimethylsilylethynylbenzene, trimethyl 4-chlorophenylethynyl silane, 2 4'-chlorophenyl-1-trimethylsilylethyne | |
2-(4-chlorophenyl)ethynyl-trimethylsilane |
Specifications
47°C to 51°C | |
Brown to White | |
C11H13ClSi | |
104°C (219°F) | |
HVIHXUCWKNRJTC-UHFFFAOYSA-N | |
2-(4-chlorophenyl)ethynyl-trimethylsilane | |
3337298 | |
97% |
78704-49-1 | |
Solid | |
MFCD03427267 | |
4-chlorophenylethynyl trimethylsilane, 4-chlorophenyl ethynyl trimethylsilane, 2-4-chlorophenyl ethynyl trimethylsilane, 2 4-chlorophenyl-1-trimethylsilylethyne, 2-4-chlorophenyl ethynyl-trimethylsilane, acmc-1bg9d, chloro-4-trimethylsilylethynylbenzene, trimethyl 4-chlorophenylethynyl silane, 2 4'-chlorophenyl-1-trimethylsilylethyne | |
C[Si](C)(C)C#CC1=CC=C(C=C1)Cl | |
208.76 | |
208.76 | |
(4-Chlorophenylethynyl)trimethylsilane |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures