missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylethynyl)trimethylsilane, 97%, Thermo Scientific™

Product Code. 15471278
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78704-49-1
Molecular Formula C11H13ClSi
Molecular Weight (g/mol) 208.76
MDL Number MFCD03427267
InChI Key HVIHXUCWKNRJTC-UHFFFAOYSA-N
Synonym 4-chlorophenylethynyl trimethylsilane, 4-chlorophenyl ethynyl trimethylsilane, 2-4-chlorophenyl ethynyl trimethylsilane, 2 4-chlorophenyl-1-trimethylsilylethyne, 2-4-chlorophenyl ethynyl-trimethylsilane, acmc-1bg9d, chloro-4-trimethylsilylethynylbenzene, trimethyl 4-chlorophenylethynyl silane, 2 4'-chlorophenyl-1-trimethylsilylethyne
PubChem CID 3337298
IUPAC Name 2-(4-chlorophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=C(C=C1)Cl

Specifications

Melting Point 47°C to 51°C
Color Brown to White
Physical Form Solid
Quantity 5 g
Flash Point 104°C (219°F)
Formula Weight 208.76
Percent Purity 97%
Chemical Name or Material (4-Chlorophenylethynyl)trimethylsilane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'