Learn More
4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals
£57.80 - £367.00
Chemical Identifiers
CAS | 873-77-8, 109-99-9, 108-88-3 |
---|---|
Molecular Formula | C6H4BrClMg |
Molecular Weight (g/mol) | 215.76 |
MDL Number | MFCD00009926 |
InChI Key | CDEMHJCJMMOFMB-UHFFFAOYSA-M |
Synonym | 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent |
PubChem CID | 101888 |
SMILES | ClC1=CC=C([Mg]Br)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10699414
|
Thermo Scientific Acros
377251000 |
100 mL | AcroSeal™ Glass bottle |
£57.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10566753
|
Thermo Scientific Acros
377258000 |
800 mL | AcroSeal™ Glass Bottle |
£367.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
873-77-8, 109-99-9, 108-88-3 | |
215.76 | |
CDEMHJCJMMOFMB-UHFFFAOYSA-M | |
101888 |
C6H4BrClMg | |
MFCD00009926 | |
4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent | |
ClC1=CC=C([Mg]Br)C=C1 |
Specifications
Yellowish-green solution | |
21.58, 76.27 | |
C6H4BrClMg | |
MFCD00009926 | |
1.0000g/mL | |
Solubility in water: vigorous reaction. | |
ClC1=CC=C([Mg]Br)C=C1 | |
215.76 | |
215.76 | |
−21°C |
873-77-8, 109-99-9, 108-88-3 | |
23.73, 78.42 | |
Cl(C6H6)MgBr | |
1M, exact strength on the certificate of analysis | |
4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent | |
CDEMHJCJMMOFMB-UHFFFAOYSA-M | |
bromo(4-chlorophenyl)magnesium | |
101888 | |
1 | |
4-Chlorophenylmagnesium bromide |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Suspected of damaging the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
- 4-Chlorophenylmagnesium bromide, 0.9M solution in THF/toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only