missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylsulfonyl)acetonitrile, 98+%, Thermo Scientific™

Product Code. 11471858
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(4-Chlorophenylsulfonyl)acetonitrile react with trimethoxymethyl-benzene, and gives 2-(4-chloro-benzenesulfonyl)-3-methoxy-3-phenyl-acrylonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1851-09-8
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.651
MDL Number MFCD00045626
InChI Key HAQGVGPNKGGSMK-UHFFFAOYSA-N
Synonym 4-chlorophenylsulfonylacetonitrile, 2-4-chlorophenyl sulfonyl acetonitrile, 4-chlorophenyl cyanomethyl sulfone, 2-4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonylacetonitrile, 4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonyl acetonitrile, 4-chlorophenyl sulfonyl acetonitrile, 2-4-chlorophenyl sulfonylacetonitrile, 4-chlorophenylsulfonyl acetonitrile
PubChem CID 735829
IUPAC Name 2-(4-chlorophenyl)sulfonylacetonitrile
SMILES C1=CC(=CC=C1S(=O)(=O)CC#N)Cl

Specifications

Melting Point 169°C to 173°C
Quantity 25 g
UN Number UN3439
Beilstein 1819678
Solubility Information Insoluble in water.
Formula Weight 215.66
Percent Purity ≥98%
Chemical Name or Material (4-Chlorophenylsulfonyl)acetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'