missing translation for 'onlineSavingsMsg'
Learn More

4-(Chlorosulfonyl)benzoic acid, 96%

Product Code. 11413577
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11413577

Brand: Thermo Scientific Alfa Aesar A17337.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10130-89-9 | C7H5ClO4S | 220.623 g/mol

It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10130-89-9
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.623
MDL Number MFCD00007448
InChI Key PTCSSXYPZOFISK-UHFFFAOYSA-N
Synonym 4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
PubChem CID 82385
IUPAC Name 4-chlorosulfonylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl

Specifications

Melting Point 228°C to 232°C
Odor Acrid
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 220.63
Percent Purity 96%
Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.