missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[3,2-d]pyrimidine, 97%, Thermo Scientific™

Product Code. 10533032
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 16269-66-2
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.614
MDL Number MFCD00122144
InChI Key TWTODSLDHCDLDR-UHFFFAOYSA-N
Synonym 4-chlorothieno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 4-chloro, 4-chloro-thieno 3,2-d pyrimidine, 4-chlorothieno 3,2-d-pyrimidine, 4-chlorothiopheno 3,2-d pyrimidine, pubchem14662, 4-chlorothienopyrimidine, acmc-209dov
PubChem CID 2735821
IUPAC Name 4-chlorothieno[3,2-d]pyrimidine
SMILES C1=CSC2=C1N=CN=C2Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.62
Percent Purity 97%
Chemical Name or Material 4-Chlorothieno[3, 2-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'