missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanone, 96%, Thermo Scientific Chemicals

Product Code. 10295860
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 491-37-2
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00006840
InChI Key MSTDXOZUKAQDRL-UHFFFAOYSA-N
Synonym 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone
PubChem CID 68110
IUPAC Name 2,3-dihydrochromen-4-one
SMILES C1COC2=CC=CC=C2C1=O

Spezifikation

CAS Min % 95.0
CAS Max % 100.0
Melting Point 34°C to 40°C
Color White to Yellow
Boiling Point 127°C to 128°C (13.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.575
Quantity 10 g
Beilstein 17, II, 335
Formula Weight 148.16
Percent Purity 96%
Physical Form Low Melting Mass, Crystals or Crystalline Powder
Chemical Name or Material 4-Chromanone, 96%
Mehr anzeigen Weniger anzeigen
Produktidentifikator
  • 4-Chromanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'