Learn More
4-Chromanone, 98+%, Thermo Scientific Chemicals
£29.90 - £326.00
Chemical Identifiers
CAS | 491-37-2 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.161 |
MDL Number | MFCD00006840 |
InChI Key | MSTDXOZUKAQDRL-UHFFFAOYSA-N |
Synonym | 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone |
PubChem CID | 68110 |
IUPAC Name | 2,3-dihydrochromen-4-one |
SMILES | C1COC2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413584
|
Thermo Scientific Alfa Aesar
A11242.06 |
5 g |
£29.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423584
|
Thermo Scientific Alfa Aesar
A11242.14 |
25 g |
£100.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433584
|
Thermo Scientific Alfa Aesar
A11242.22 |
100 g |
£326.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and moisture. Store away from oxidizing agents. Store at 2-8°C.
Chemical Identifiers
491-37-2 | |
148.161 | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
68110 | |
C1COC2=CC=CC=C2C1=O |
C9H8O2 | |
MFCD00006840 | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
2,3-dihydrochromen-4-one |
Specifications
491-37-2 | |
127°C to 128°C (13 mmHg) | |
C9H8O2 | |
MFCD00006840 | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
2,3-dihydrochromen-4-one | |
68110 | |
≥98% |
36°C to 40°C | |
110°C (230°F) | |
1.575 | |
124652 | |
Slightly soluble in water. | |
C1COC2=CC=CC=C2C1=O | |
148.161 | |
148.16 | |
4-Chromanone |
Safety and Handling
missing translation for 'einecsNumber' : 207-736-3
missing translation for 'rtecsNumber' : DJ2981225
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only