missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4-(4-fluorophenyl)cyclohexanone, 98%, Thermo Scientific™

Product Code. 15493647
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56326-98-8
Molecular Formula C13H12FNO
Molecular Weight (g/mol) 217.24
MDL Number MFCD00044815
InChI Key FNWWTNNFZGBMEM-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-4-oxocyclohexanecarbonitrile, 4-cyano-4-4-fluorophenyl cyclohexanone, 1-4-fluorophenyl-4-oxocyclohexane-1-carbonitrile, cyclohexanecarbonitrile, 1-4-fluorophenyl-4-oxo, acmc-1b1x4, dsstox_cid_31566, dsstox_rid_97451, dsstox_gsid_57777, 4-cyano-4-p-fluorophenylcyclohexanone, 4-cyano-4-p-fluorophenyl cyclohexanone
PubChem CID 91805
IUPAC Name 1-(4-fluorophenyl)-4-oxocyclohexane-1-carbonitrile
SMILES FC1=CC=C(C=C1)C1(CCC(=O)CC1)C#N

Specifications

Melting Point 91°C to 94°C
Quantity 250 mg
UN Number UN3439
Formula Weight 217.25
Percent Purity 98%
Chemical Name or Material 4-Cyano-4-(4-fluorophenyl)cyclohexanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'