missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-heptyloxybiphenyl, 99%, Thermo Scientific™

Product Code. 11449537
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52364-72-4
Molecular Formula C20H23NO
Molecular Weight (g/mol) 293.41
MDL Number MFCD00134146
InChI Key JPBFKTCKZLMJED-UHFFFAOYSA-N
Synonym 4'-heptyloxy-4-cyanobiphenyl, 4-cyano-4'-heptyloxybiphenyl, 4'-heptyloxy-1,1'-biphenyl-4-carbonitrile, 4'-heptyloxy biphenyl-4-carbonitrile, 7cob, 1,1'-biphenyl-4-carbonitrile, 4'-heptyloxy, 4-heptyloxycyanodiphenyl, 4-cyano-4'-heptoxybiphenyl, 4-4-heptoxyphenyl benzonitrile, 4-4-heptyloxyphenyl benzonitrile
PubChem CID 104172
IUPAC Name 4-(4-heptoxyphenyl)benzonitrile
SMILES CCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 53°C to 55°C
Quantity 5 g
Formula Weight 293.41
Percent Purity 99%
Chemical Name or Material 4-Cyano-4'-n-heptyloxybiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'