missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-hexyloxybiphenyl, 97%, Thermo Scientific™

Product Code. 15412268
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Cyano-4′-n-hexyloxybiphenyl, is used as a intermediate of liquid crystals. It is mainly used as the pharmaceutical intermediate, liquid crystal intermediate, liquid crystal monomers.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41424-11-7
Molecular Formula C19H21NO
Molecular Weight (g/mol) 279.38
MDL Number MFCD00134145
InChI Key YUYXUPYNSOFWFV-UHFFFAOYSA-N
Synonym 4'-hexyloxy-4-biphenylcarbonitrile, 6ocb, liquid crystal 549, 1,1'-biphenyl-4-carbonitrile, 4'-hexyloxy, 4-hexyloxycyanodiphenyl, 4-cyano-4'-hexoxybiphenyl, 4-4-hexyloxyphenyl benzonitrile, 4'-hexyloxy-1,1'-biphenyl-4-carbonitrile
PubChem CID 162465
IUPAC Name 4-(4-hexoxyphenyl)benzonitrile
SMILES CCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 279.38
Percent Purity 97%
Chemical Name or Material 4-Cyano-4'-n-hexyloxybiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'