missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-4'-n-pentylbiphenyl, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Cyano-4'-n-pentylbiphenyl is used in the preparation of 4'-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyano-4′-n-pentylbiphenyl is used in the preparation of 4′-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.
Solubility
Miscible with Acetone, chloroform, toluene, acetonitrile, methylene chloride. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
4-Cyano-4′-n-pentylbiphenyl is used in the preparation of 4′-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.
Solubility
Miscible with Acetone, chloroform, toluene, acetonitrile, methylene chloride. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 22°C to 23°C |
Density | 1.008 |
Boiling Point | 212°C to 215°C (3 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.533 |
Quantity | 5 g |
UN Number | UN3276 |
Solubility Information | Miscible with Acetone,chloroform,toluene,acetonitrile,methylene chloride. Immiscible with water. |
Formula Weight | 249.36 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'