missing translation for 'onlineSavingsMsg'
Learn More
4-Cyano-4-phenylpiperidine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 11452040

4-Cyano-4-phenylpiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11452040
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11452040

Brand: Thermo Scientific Alfa Aesar H28414.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51304-58-6
Molecular Formula C12H15ClN2
Molecular Weight (g/mol) 222.716
MDL Number MFCD00012775
InChI Key CQPHZBOPSZGTJM-UHFFFAOYSA-N
Synonym 4-cyano-4-phenylpiperidine hydrochloride, 4-phenylpiperidine-4-carbonitrile hydrochloride, 4-cyano-4-phenylpiperidinehydrochloride, 4-phenylpiperidine-4-carbonitrile, chloride, 4-phenylpiperidine-4-carbonitrile hcl, pubchem19077, acmc-20a3zw, 4-cyano-4-phenylpiperidine hcl, timtec-bb sbb003334, 4-phenylisonipecotonitrile hydrochlorideShow MoreShow Less
PubChem CID 3084973
IUPAC Name 4-phenylpiperidine-4-carbonitrile;hydrochloride
SMILES C1CNCCC1(C#N)C2=CC=CC=C2.Cl

Specifications

Melting Point 210°C to 214°C
Quantity 1 g
UN Number UN3439
Formula Weight 222.71
Percent Purity 97%
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243661 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberH28414.09,H28414.03
Lot Number10154845 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28414.09,H28414.03
Lot Number10237045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28414.09,H28414.03
Lot Number10203353 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28414.09,H28414.03
Lot NumberE27U036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH28414.09,H28414.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Cyano-4-phenylpiperidine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.