missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzophenone, 98%, Thermo Scientific Chemicals

Product Code. 11402428
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1503-49-7
Molecular Formula C14H9NO
Molecular Weight (g/mol) 207.232
MDL Number MFCD00016382
InChI Key YSZWJJANSNFQMM-UHFFFAOYSA-N
PubChem CID 73921
IUPAC Name 4-benzoylbenzonitrile
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
UN Number UN3439
Beilstein 641181
Synonym 4-cyanobenzophenone, 4-benzoyl-benzonitrile, benzonitrile, 4-benzoyl, 4-cyano-benzophenon, 4-cyano-benzophenone, acmc-20amk8, #, benzonitrile,4-benzoyl, 4-phenylcarbonyl benzenecarbonitrile
Solubility Information Insoluble in water.
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'