missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobutylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as primary and secondary intermediate.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids a
It is used as primary and secondary intermediate.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids a
Specifications
CAS | 226570-68-9 |
Density | 0.971 |
Molecular Formula | C5H9BrNZn- |
Linear Formula | N≡C(CH2)4ZnBr |
MDL Number | MFCD01316996 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive |
Synonym | 4-cyanobutylzinc bromide 0.5m in thf |
Solubility Information | Reacts with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'