missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanoindole, 97%, Thermo Scientific™
£32.90 - £93.51
Chemical Identifiers
CAS | 16136-52-0 |
---|---|
Molecular Formula | C9H6N2 |
Molecular Weight (g/mol) | 142.16 |
InChI Key | CEUFGDDOMXCXFW-UHFFFAOYSA-N |
Synonym | 4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide |
PubChem CID | 3817602 |
IUPAC Name | 1H-indole-4-carbonitrile |
SMILES | C1=CC(=C2C=CNC2=C1)C#N |
Description
Chemical Identifiers
16136-52-0 | |
142.16 | |
4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide | |
1H-indole-4-carbonitrile |
C9H6N2 | |
CEUFGDDOMXCXFW-UHFFFAOYSA-N | |
3817602 | |
C1=CC(=C2C=CNC2=C1)C#N |
Specifications
16136-52-0 | |
100.0 | |
White to Brown | |
96% min. (HPLC) | |
C9H6N2 | |
CEUFGDDOMXCXFW-UHFFFAOYSA-N | |
1H-indole-4-carbonitrile | |
3817602 | |
97% | |
4-Cyanoindole |
96.0 | |
120°C to 121°C | |
Authentic | |
Glass Bottle | |
4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide | |
C1=CC(=C2C=CNC2=C1)C#N | |
142.16 | |
142.16 | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Cyanoindole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only