Learn More
4-Cyanophenyl isocyanate, 95%, Thermo Scientific Chemicals
£92.90 - £345.00
Chemical Identifiers
CAS | 40465-45-0 |
---|---|
Molecular Formula | C8H4N2O |
Molecular Weight (g/mol) | 144.13 |
MDL Number | MFCD00037038 |
InChI Key | TVSXDZNUTPLDKY-UHFFFAOYSA-N |
PubChem CID | 170380 |
IUPAC Name | 4-isocyanatobenzonitrile |
SMILES | C1=CC(=CC=C1C#N)N=C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10450663
|
Thermo Scientific Acros
405970050 |
5 g | Glass bottle |
£92.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10258473
|
Thermo Scientific Acros
405970250 |
25 g | Glass bottle |
£345.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40465-45-0 | |
144.13 | |
TVSXDZNUTPLDKY-UHFFFAOYSA-N | |
4-isocyanatobenzonitrile |
C8H4N2O | |
MFCD00037038 | |
170380 | |
C1=CC(=CC=C1C#N)N=C=O |
Specifications
40465-45-0 | |
100.0 | |
White to Beige | |
94% min. (GC) | |
NCC6H4NCO | |
4-cyanophenyl isocyanate, benzonitrile, 4-isocyanato, 4-cyanophenylisocyanate, p-cyanophenyl isocyanate, benzonitrile,4-isocyanato, 4-isocyanatobenzenecarbonitrile, 4-cyanophenylisocyanat, p-cyanophenylisocyanate, #, 1-cyano-4-isocyanato-benzene | |
TVSXDZNUTPLDKY-UHFFFAOYSA-N | |
4-isocyanatobenzonitrile | |
170380 | |
95% | |
4-Cyanophenyl isocyanate |
94.0 | |
102°C | |
Authentic | |
C8H4N2O | |
MFCD00037038 | |
Solubility in water: insoluble (dec). Other solubilities: soluble in toluene and chloroform | |
C1=CC(=CC=C1C#N)N=C=O | |
144.13 | |
144.13 | |
Crystalline Needles or Solid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 254-934-
- 4-Cyanophenyl isocyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only