missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15465335
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanophenylzinc bromide is used to produce biphenyl-4-carbonitrile.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents, air, bases and heat. Air Sensitive. Store under dry iner
TRUSTED_SUSTAINABILITY

Specifications

CAS 131379-14-1
Density 0.98
Molecular Formula C7H4BrNZn
MDL Number MFCD00671984
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym zinc 2+ ion 4-cyanobenzen-1-ide bromide, zinc bromide 4-cyanobenzen-1-ide 1/1/1
Solubility Information Reacts with water.
InChI Key HEOMKTNNZDZMJZ-UHFFFAOYSA-M
SMILES C1=CC(=CC=[C-]1)C#N.[Zn+2].[Br-]
IUPAC Name zinc;benzonitrile;bromide
Molecular Weight (g/mol) 247.4
PubChem CID 54750729
Formula Weight 247.4
Chemical Name or Material 4-Cyanophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'