missing translation for 'onlineSavingsMsg'
Learn More

4-(Di-n-propylsulfamoyl)benzoyl chloride, 96%, Thermo Scientific™

Product Code. 11460260
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29171-72-0
Molecular Formula C13H18ClNO3S
Molecular Weight (g/mol) 303.80
MDL Number MFCD00121817
InChI Key LCQXGFTXKWEWQC-UHFFFAOYSA-N
Synonym 4-dipropylsulfamoyl benzoyl chloride, 4-dipropylamino sulfonyl benzene-1-carbonyl chloride, 4-n,n-dipropylsulfamoyl benzoyl chloride, benzoyl chloride,4-dipropylamino sulfonyl, acmc-1cejw, 4-dipropylsulfamoyl-benzoyl chloride, 4-di-n-propylsulfamoyl benzoyl chloride, 4-dipropylaminosulfonyl benzoyl chloride
PubChem CID 2735528
IUPAC Name 4-(dipropylsulfamoyl)benzoyl chloride
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 59°C to 61°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 303.81
Percent Purity 96%
Chemical Name or Material 4-(Di-n-propylsulfamoyl)benzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.