missing translation for 'onlineSavingsMsg'
Learn More
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals Product Code.: 11436776

4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals

Product Code. 11436776
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11436776

Brand: Thermo Scientific Alfa Aesar B23915.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2481-94-9
Molecular Formula C16H19N3
Molecular Weight (g/mol) 253.349
MDL Number MFCD00009043
InChI Key SJJISKLXUJVZOA-UHFFFAOYSA-N
Synonym 4-diethylamino azobenzene, oil yellow de, diethyl yellow, c.i. solvent yellow 56, solvent yellow 56, 4-diethylaminoazobenzene, oil yellow ga, oil yellow nb, oil yellow dea, oil yellow encShow MoreShow Less
PubChem CID 17204
IUPAC Name N,N-diethyl-4-phenyldiazenylaniline
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=CC=C2

Specifications

Melting Point 93°C to 96°C
Quantity 25 g
Formula Weight 253.35
Percent Purity 98%
Chemical Name or Material 4-(Diethylamino)azobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178844 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog NumberB23915.06,B23915.22,B23915.14
Lot Number10244568 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberB23915.06,B23915.22,B23915.14
Lot Number10173626 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23915.06,B23915.22,B23915.14
Lot NumberG6883A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23915.06,B23915.22,B23915.14
Lot Number10160184 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23915.06,B23915.22,B23915.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.