missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)-2-fluorobenzonitrile, 97%, Thermo Scientific™

Product Code. 15424307
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-49-5
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD09832366
InChI Key OERIHGQAGTVWPN-UHFFFAOYSA-N
Synonym 4-difluoromethoxy-2-fluorobenzonitrile, 4-difluoromethoxy-2-fluoro-benzonitrile, 4-difluoromethoxy-2-fluorobenzenecarbonitrile, 4-difluoromethoxy-2-fluorobenzonitrile, jrd
PubChem CID 20111957
IUPAC Name 4-(difluoromethoxy)-2-fluorobenzonitrile
SMILES C1=CC(=C(C=C1OC(F)F)F)C#N

Specifications

Melting Point 41°C to 43°C
Quantity 1 g
UN Number UN3439
Formula Weight 187.12
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)-2-fluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'