missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™

Product Code. 15440086
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878760-47-5
Molecular Formula C8H9F3N2O2
Molecular Weight (g/mol) 222.167
MDL Number MFCD05662688
InChI Key CHSZUWDVGYPDPG-UHFFFAOYSA-N
Synonym 4-dimethoxymethyl-2-trifluoromethyl pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl-pyrimidine, pyrimidine,4-dimethoxymethyl-2-trifluoromethyl, dimethoxy 2-trifluoromethyl pyrimidin-4-yl methane, pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl
PubChem CID 17750125
IUPAC Name 4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine
SMILES COC(C1=NC(=NC=C1)C(F)(F)F)OC

Specifications

Quantity 1 g
Formula Weight 222.17
Percent Purity 97%
Chemical Name or Material 4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'