missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)benzaldehyde, ≥99% (HPLC), Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 73°C to 75°C |
Color | White |
Boiling Point | 176°C to 177°C |
Flash Point | 164°C (327.2°F) |
Ignition Residue | ≤0.1% (as SO4) |
Loss on Drying | 0 to 0.5% |
Packaging | Glass Bottle |
Linear Formula | (CH3)2NC6H4CHO |
Quantity | 250 g |
UN Number | NONH for all modes of transport |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'