missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzonitrile, 95%, Thermo Scientific Chemicals

£24.90 - £90.60

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001815
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10254140
View Documents
Thermo Scientific Acros
115780050
5 g Glass bottle
£24.90
5g
Please to purchase this item. Need a web account? Register with us today!
10756141
View Documents
Thermo Scientific Acros
115780250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1197-19-9
146.19
JYMNQRQQBJIMCV-UHFFFAOYSA-N
70967
CN(C)C1=CC=C(C=C1)C#N
C9H10N2
MFCD00001815
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
4-(dimethylamino)benzonitrile

Specifications

1197-19-9
100.0
Brown
Authentic
C9H10N2
MFCD00001815
4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
CN(C)C1=CC=C(C=C1)C#N
146.19
146.19
Crystalline Powder
94.0
70.0°C to 76.0°C
318.0°C
95%
(CH3)2NC6H4CN
14, 428
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-(dimethylamino)benzonitrile
70967
95%
4-(Dimethylamino)benzonitrile, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-819-8

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only