missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)pyridine tribromide, 97%, Thermo Scientific Chemicals

Product Code. 15401128
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92976-81-3
Molecular Formula C7H10Br3N2-
Molecular Weight (g/mol) 361.883
MDL Number MFCD00043439
InChI Key TYDZHQMKSLAFKM-UHFFFAOYSA-N
Synonym 4-dimethylamino pyridine hydrobromide perbromide
PubChem CID 131863682
SMILES CN(C)C1=CC=NC=C1.Br[Br-]Br

Specifications

Melting Point 129°C to 132°C
Quantity 25 g
UN Number UN3261
Formula Weight 362.89
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)pyridine tribromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.