missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, 99+%, Thermo Scientific Chemicals

Artikelnummer. 11905321
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Spezifikation

Melting Point 70°C to 75°C
Color Green to Yellow
Boiling Point 176°C to 177°C (17.0 mmHg)
Flash Point 164°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 2.5 kg
Beilstein 14,31
Fieser 01,273; 02,146
Merck Index 15,3252
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in alcohol,ether and chloroform
Formula Weight 149.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'