missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylaminobenzaldehyde, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
25 g
Specifications
Melting Point | 70°C |
Color | Yellow |
Boiling Point | 176°C |
Packaging | Plastic powder jar |
Quantity | 25 g |
Formula Weight | 149.19g/mol |
Physical Form | Powder |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 4-Dimethylaminobenzaldehyde
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'