missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminocinnamaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11406446
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)cinnamaldehyde is a reagent used in the assay of apotryptophanase and tryptophanase. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

Solubility
Soluble in chloroform, ethanol, dioxane.

Notes
Air & light sensitive. Store at -20°C. Store in dark. Incompatible with oxidizing agents, air, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6203-18-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00007002
InChI Key RUKJCCIJLIMGEP-ONEGZZNKSA-N
Synonym 4-dimethylamino cinnamaldehyde, 4-dimethylaminocinnamaldehyde, p-dimethylaminocinnamaldehyde, 3-4-dimethylamino phenyl acrylaldehyde, 4-dimethylcinnamaldehyde, p-dimethylaminocinnamic aldehyde, p-dimethylamino cinnamaldehyde, cinnamaldehyde, p-dimethylamino, unii-9rsi7wz9f0, dmaca reagent
PubChem CID 5284506
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enal
SMILES CN(C)C1=CC=C(C=C1)C=CC=O

Specifications

Melting Point 137°C to 144°C
Quantity 5 g
Beilstein 972369
Sensitivity Air and light sensitive
Solubility Information Soluble in chloroform,ethanol,dioxane.
Formula Weight 175.23
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminocinnamaldehyde
compliance-icons
Product Identifier
  • p-Dimethylaminocinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : p-Dimethylaminocinnamaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'