missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 99% |
Packaging | Glass Bottle |
Molecular Formula | C7H10N2 |
Linear Formula | (C5H4N)N(CH3)2 |
Quantity | 100 g |
Beilstein | 22, II, 341 |
Fieser | 03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142 |
Merck Index | 15, 3433 |
Show More |
Safety and Handling




Product Identifier
- 4-Dimethylaminopyridine
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction